Assay Details
| Gene Symbol: Gjb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010291.3 | NP_034421.1 | 1 - 2 | 140 | 93 | |
| GenBank mRNA | AK082917.1 | - | 1 - 2 | 141 | 93 | |
| M91236.1 | - | 1 - 2 | 97 | 93 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK082917.1,M91236.1,NM_010291.3 transcripts.
Set Membership: |
|

Gjb5
Genomic Map