Assay Details
| Gene Symbol: Gzma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010370.2 | NP_034500.1 | 1 - 2 | 101 | 77 | |
| GenBank mRNA | AK146132.1 | - | 1 - 2 | 98 | 77 | |
| AK156728.1 | - | 1 - 2 | 100 | 77 | ||
| BC061146.1 | - | 1 - 2 | 101 | 30296854 | 77 | |
| M26183.1 | - | 1 - 2 | 89 | 77 | ||
| X14799.1 | - | 1 - 2 | 103 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gzma
Genomic Map