Assay Details
| Gene Symbol: Hc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010406.2 | NP_034536.1 | 31 - 32 | 4140 | 91 | |
| GenBank mRNA | BC003963.1 | - | 2 - 3 | 184 | 91 | |
| M35525.1 | - | 31 - 32 | 4095 | 91 | ||
| M35526.1 | - | 31 - 32 | 4093 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hc
Genomic Map