Assay Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010484.2 | NP_034614.2 | 6 - 7 | 1160 | 56 | |
GenBank mRNA | AF013604.1 | - | 5 - 6 | 973 | 56 | |
BC108978.1 | - | 6 - 7 | 1142 | 40047534 | 56 | |
BC108979.1 | - | 6 - 7 | 1142 | 40047537 | 56 | |
X66119.1 | - | 5 - 6 | 754 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
