Assay Details
Gene Symbol: Mmp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008611.4 | NP_032637.3 | 1 - 2 | 213 | 86 | |
GenBank mRNA | AK089234.1 | - | 1 - 2 | 184 | 86 | |
AK089336.1 | - | 1 - 2 | 187 | 86 | ||
AK137468.1 | - | 1 - 2 | 185 | 86 | ||
AK154937.1 | - | 1 - 2 | 184 | 86 | ||
AK155740.1 | - | 1 - 2 | 184 | 86 | ||
AK155832.1 | - | 1 - 2 | 184 | 86 | ||
AK155901.1 | - | 1 - 2 | 187 | 86 | ||
AK170177.1 | - | 1 - 2 | 184 | 86 | ||
AK170760.1 | - | 1 - 2 | 184 | 86 | ||
AK171168.1 | - | 1 - 2 | 184 | 86 | ||
AK171613.1 | - | 1 - 2 | 186 | 86 | ||
BC042742.1 | - | 1 - 2 | 194 | 5032770 | 86 | |
BY204369.1 | - | 1 - 2 | 214 | 86 | ||
U96696.1 | - | 1 - 2 | 179 | 86 | ||
Y13342.1 | - | 1 - 2 | 111 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
