Assay Details
| Gene Symbol: Igf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122736.1 | NP_001116208.1 | 1 - 2 | 100 | 99 | |
| GenBank mRNA | CN460564.1 | - | 1 - 2 | 312 | 99 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001122736.1 transcripts.
Set Membership: |
|

Igf2
Genomic Map