Assay Details
Gene Symbol: Il4ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008700.3 | NP_001008700.1 | 4 - 5 | 450 | 71 | |
GenBank mRNA | AK034466.1 | - | 4 - 5 | 494 | 71 | |
AK042342.1 | - | 4 - 5 | 450 | 71 | ||
AK088086.1 | - | 4 - 5 | 462 | 71 | ||
AK137827.1 | - | 4 - 5 | 462 | 71 | ||
AK149603.1 | - | 4 - 5 | 463 | 71 | ||
AK149717.1 | - | 4 - 5 | 463 | 71 | ||
AK150584.1 | - | 4 - 5 | 462 | 71 | ||
AK151998.1 | - | 4 - 5 | 462 | 71 | ||
AK153186.1 | - | 4 - 5 | 462 | 71 | ||
AK159375.1 | - | 4 - 5 | 461 | 71 | ||
AK160016.1 | - | 4 - 5 | 454 | 71 | ||
BC112911.1 | - | 4 - 5 | 450 | 30138743 | 71 | |
BC128301.1 | - | 3 - 4 | 321 | 71 | ||
BC128302.1 | - | 3 - 4 | 321 | 71 | ||
KC212685.1 | - | 342 | 71 | |||
M27959.1 | - | 4 - 5 | 445 | 71 | ||
M27960.1 | - | 4 - 5 | 445 | 71 | ||
M29854.1 | - | 4 - 5 | 429 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
