Assay Details
| Gene Symbol: Itk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281965.1 | NP_001268894.1 | 10 - 11 | 1111 | 109 | |
| NM_001281966.1 | NP_001268895.1 | 10 - 11 | 1102 | 109 | ||
| NM_001281968.1 | NP_001268897.1 | 10 - 11 | 1090 | 109 | ||
| NM_010583.3 | NP_034713.2 | 10 - 11 | 1093 | 109 | ||
| GenBank mRNA | AK037554.1 | - | 173 | 109 | ||
| AK083380.1 | - | 10 - 11 | 1065 | 109 | ||
| AK153613.1 | - | 10 - 11 | 1049 | 109 | ||
| BC128374.1 | - | 10 - 11 | 1019 | 40111687 | 109 | |
| BC128375.1 | - | 10 - 11 | 1016 | 40111688 | 109 | |
| D14042.1 | - | 10 - 11 | 1033 | 109 | ||
| L00619.1 | - | 10 - 11 | 1098 | 109 | ||
| L05631.1 | - | 10 - 11 | 1059 | 109 | ||
| L10628.1 | - | 10 - 11 | 1061 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itk
Genomic Map