Assay Details
Gene Symbol: Kcnab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289450.1 | NP_001276379.1 | 5 - 6 | 373 | 98 | |
NM_010597.4 | NP_034727.3 | 4 - 5 | 789 | 98 | ||
GenBank mRNA | AF033003.1 | - | 4 - 5 | 1144 | 98 | |
AK004666.1 | - | 5 - 6 | 357 | 98 | ||
AK015412.1 | - | 5 - 6 | 629 | 98 | ||
AK031362.1 | - | 4 - 5 | 790 | 98 | ||
AK138467.1 | - | 4 - 5 | 778 | 98 | ||
AK153200.1 | - | 5 - 6 | 375 | 98 | ||
BC014701.1 | - | 4 - 5 | 514 | 4482268 | 98 | |
U65591.1 | - | 4 - 5 | 385 | 98 | ||
X97281.1 | - | 4 - 5 | 773 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
