Assay Details
Gene Symbol: Kcnab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289450.1 | NP_001276379.1 | 12 - 13 | 894 | 83 | |
NM_010597.4 | NP_034727.3 | 11 - 12 | 1310 | 83 | ||
GenBank mRNA | AF033003.1 | - | 11 - 12 | 1665 | 83 | |
AK004666.1 | - | 12 - 13 | 878 | 83 | ||
AK015412.1 | - | 13 - 14 | 1150 | 83 | ||
AK031362.1 | - | 11 - 12 | 1311 | 83 | ||
AK138467.1 | - | 11 - 12 | 1299 | 83 | ||
AK153200.1 | - | 12 - 13 | 896 | 83 | ||
BC014701.1 | - | 11 - 12 | 1035 | 4482268 | 83 | |
U65591.1 | - | 11 - 12 | 906 | 83 | ||
X97281.1 | - | 11 - 12 | 1294 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
