Assay Details
| Gene Symbol: Matk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285853.1 | NP_001272782.1 | 5 - 6 | 784 | 87 | |
| NM_001285854.1 | NP_001272783.1 | 5 - 6 | 781 | 87 | ||
| NM_001285855.1 | NP_001272784.1 | 5 - 6 | 849 | 87 | ||
| NM_010768.2 | NP_034898.1 | 6 - 7 | 917 | 87 | ||
| GenBank mRNA | AK013606.1 | - | 5 - 6 | 598 | 87 | |
| AK018182.1 | - | 5 - 6 | 635 | 87 | ||
| BC111864.1 | - | 5 - 6 | 595 | 40046807 | 87 | |
| D45243.1 | - | 5 - 6 | 849 | 87 | ||
| L27738.1 | - | 6 - 7 | 671 | 87 | ||
| S77473.1 | - | 5 - 6 | 596 | 87 | ||
| U05210.1 | - | 5 - 6 | 570 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Matk
Genomic Map