Assay Details
| Gene Symbol: Matk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285853.1 | NP_001272782.1 | 10 - 11 | 1201 | 72 | |
| NM_001285854.1 | NP_001272783.1 | 10 - 11 | 1198 | 72 | ||
| NM_001285855.1 | NP_001272784.1 | 10 - 11 | 1266 | 72 | ||
| NM_010768.2 | NP_034898.1 | 11 - 12 | 1334 | 72 | ||
| GenBank mRNA | AK013606.1 | - | 10 - 11 | 1015 | 72 | |
| D45243.1 | - | 10 - 11 | 1266 | 72 | ||
| L27738.1 | - | 11 - 12 | 1088 | 72 | ||
| S77473.1 | - | 10 - 11 | 1013 | 72 | ||
| U05210.1 | - | 10 - 11 | 987 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Matk
Genomic Map