Assay Details
| Gene Symbol: Mmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010809.1 | NP_034939.1 | 7 - 8 | 1154 | 88 | |
| GenBank mRNA | AK148467.1 | - | 7 - 8 | 1132 | 88 | |
| BC006725.1 | - | 7 - 8 | 1118 | 3962288 | 88 | |
| X63162.1 | - | 7 - 8 | 1154 | 88 | ||
| X66402.1 | - | 7 - 8 | 1096 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp3
Genomic Map