Assay Details
Gene Symbol: Sik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010831.2 | NP_034961.2 | 6 - 7 | 763 | 91 | |
GenBank mRNA | AK141817.1 | - | 6 - 7 | 763 | 91 | |
AK172427.1 | - | 6 - 7 | 93 | |||
AK220203.1 | - | 4 - 5 | 376 | 91 | ||
U11494.2 | - | 6 - 7 | 755 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
