Assay Details
Gene Symbol: Sik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010831.2 | NP_034961.2 | 8 - 9 | 1110 | 57 | |
GenBank mRNA | AK087322.1 | - | 1 - 2 | 27 | 57 | |
AK141817.1 | - | 8 - 9 | 1110 | 57 | ||
AK172427.1 | - | 8 - 9 | 1113 | 57 | ||
AK220203.1 | - | 6 - 7 | 723 | 57 | ||
U11494.2 | - | 8 - 9 | 1102 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
