Assay Details
| Gene Symbol: Sik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010831.2 | NP_034961.2 | 10 - 11 | 1385 | 77 | |
| GenBank mRNA | AK087322.1 | - | 3 - 4 | 302 | 77 | |
| AK141817.1 | - | 10 - 11 | 1385 | 77 | ||
| AK172427.1 | - | 9 - 10 | 1825 | 77 | ||
| AK220203.1 | - | 8 - 9 | 998 | 77 | ||
| U11494.2 | - | 10 - 11 | 1377 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sik1
Genomic Map