Assay Details
| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039657.2 | NP_001034746.1 | 2 - 3 | 641 | 60 | |
| NM_001039658.2 | NP_001034747.1 | 2 - 3 | 641 | 60 | ||
| NM_001286557.1 | NP_001273486.1 | 3 - 4 | 725 | 60 | ||
| NM_001286558.1 | NP_001273487.1 | 2 - 3 | 656 | 60 | ||
| GenBank mRNA | AB057423.1 | - | 2 - 3 | 629 | 60 | |
| AF329359.1 | - | 2 - 3 | 666 | 60 | ||
| AF329360.1 | - | 3 - 4 | 735 | 60 | ||
| AK015732.1 | - | 2 - 3 | 614 | 60 | ||
| AK161511.1 | - | 2 - 3 | 642 | 60 | ||
| BC024377.1 | - | 2 - 3 | 617 | 4976492 | 60 | |
| U67176.1 | - | 4 - 5 | 658 | 60 | ||
| U77383.1 | - | 2 - 3 | 278 | 60 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF329359.1,AF329360.1,NM_001286557.1,NM_001286558.1,U67176.1,U77383.1 transcripts.
Set Membership: |
|

Mtl5
Genomic Map