Assay Details
Gene Symbol: Orm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011016.2 | NP_035146.1 | 2 - 3 | 290 | 82 | |
GenBank mRNA | BC057985.1 | - | 2 - 3 | 265 | 5126236 | 82 |
CB949119.1 | - | 2 - 3 | 293 | 82 | ||
M12566.1 | - | 1 - 2 | 45 | 82 | ||
M27009.1 | - | 2 - 3 | 255 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141546 | NM_011016.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141546 | BC057985.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141546 | CB949119.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141546 | M12566.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141546 | M27009.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141546 | NM_011016.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141546 | BC057985.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141546 | CB949119.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141546 | M12566.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141546 | M27009.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471061 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539188 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163539 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141546 | NM_011016.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141546 | BC057985.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141546 | CB949119.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141546 | M12566.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141546 | M27009.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141546 | NM_011016.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141546 | BC057985.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141546 | CB949119.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141546 | M12566.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141546 | M27009.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141546 | NM_011016.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141546 | BC057985.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141546 | CB949119.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141546 | M12566.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141546 | M27009.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141546 | NM_011016.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141546 | BC057985.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141546 | CB949119.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141546 | M12566.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141546 | M27009.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22969948 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22969946 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22969943 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22969941 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22969936 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22969942 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22969932 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22969962 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22969929 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22969947 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22969953 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163539 | - | - |
Whole Mouse Genome G4122A | A_51_P141546 | NM_011016.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P141546 | BC057985.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P141546 | CB949119.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P141546 | M12566.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P141546 | M27009.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P141546 | NM_011016.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P141546 | BC057985.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P141546 | CB949119.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P141546 | M12566.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P141546 | M27009.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116125 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163539 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03335848 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03335846 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03335878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22969916 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969953 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03436921 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969932 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969941 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03335859 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03335856 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969956 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969947 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969946 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969942 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969967 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969936 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969974 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03335874 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969943 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969972 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22969948 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10505454 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505452 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505455 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505456 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505453 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420438_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505451 | - | - |
Mouse Genome 430 2.0 Array | 1420438_at | - | - |
Mouse Genome 430 2.0 Array | 1420438_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18406_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18406_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400005183.mm.1 | NM_011016.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400005183.mm.1 | NM_011016.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400005183.mm.1 | TC0400000721.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400005182.mm.1 | NM_011016.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400005182.mm.1 | NM_011016.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400005182.mm.1 | TC0400000721.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400005185.mm.1 | NM_011016.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400005185.mm.1 | NM_011016.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400005185.mm.1 | TC0400000721.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400005184.mm.1 | NM_011016.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400005184.mm.1 | NM_011016.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400005184.mm.1 | TC0400000721.mm.1 | MTA TC |
Mouse Expression Set 430 | 1420438_PM_at | - | - |
Mouse Expression Set 430 | 1420438_at | - | - |
Murine Genome U74v2 Set | 94734_at | - | - |
Murine Genome U74v2 Set | 100437_g_at | - | - |
Murine Genome U74v2 Set | 100437_PM_g_at | - | - |
Murine Genome U74v2 Set | 94734_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1420438_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1420438_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420438_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420438_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2100048 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002100048 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2591265 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2591265 | - | - |
mouse-6 v1.1 (Illumina) | scl25363.4.1_1-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2100048 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl25363.4.1_1-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100048 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2591264 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2591264 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25363.4.1_1-S | - | - |
