Assay Details
| Gene Symbol: Rorc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011281.3 | NP_035411.2 | 2 - 3 | 167 | 76 | |
| GenBank mRNA | AK160122.1 | - | 2 - 3 | 125 | 76 | |
| BC014804.1 | - | 2 - 3 | 110 | 4489193 | 76 | |
| CJ098183.1 | - | 2 - 3 | 135 | 76 | ||
| U39071.1 | - | 2 - 3 | 143 | 76 | ||
| U43508.1 | - | 2 - 3 | 105 | 76 | ||
More Information
Set Membership: |
|

Rorc
Genomic Map