Assay Details
Gene Symbol: Rps6ka2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011299.4 | NP_035429.1 | 9 - 10 | 1044 | 69 | |
GenBank mRNA | AJ131021.1 | - | 9 - 10 | 872 | 69 | |
AK019881.1 | - | 9 - 10 | 949 | 69 | ||
AK050996.1 | - | 1 - 2 | 436 | 69 | ||
BC038251.1 | - | 3 - 4 | 271 | 69 | ||
BC043064.1 | - | 9 - 10 | 994 | 5706153 | 69 | |
BC051079.1 | - | 9 - 10 | 954 | 6491001 | 69 | |
BC055331.1 | - | 9 - 10 | 1044 | 6406879 | 69 | |
BC056946.1 | - | 9 - 10 | 987 | 6822831 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
