Assay Details
| Gene Symbol: Scnn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272023.1 | NP_001258952.1 | 8 - 9 | 1373 | 69 | |
| NM_011325.2 | NP_035455.1 | 8 - 9 | 1370 | 69 | ||
| NR_073548.1 | 8 - 9 | 1370 | 69 | |||
| GenBank mRNA | AF112186.1 | - | 8 - 9 | 1297 | 69 | |
| AK028909.1 | - | 8 - 9 | 1352 | 69 | ||
| AK036819.1 | - | 8 - 9 | 1371 | 69 | ||
| AK087055.1 | - | 8 - 9 | 1332 | 69 | ||
| AK164709.1 | - | 8 - 9 | 1333 | 69 | ||
| BC131969.1 | - | 8 - 9 | 1286 | 40130246 | 69 | |
| BC132397.1 | - | 8 - 9 | 1286 | 40130674 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scnn1b
Genomic Map