Assay Details
| Gene Symbol: Clcn4-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC089497.1 | - | 1 - 2 | 321 | 77 | |
| BC110668.1 | - | 1 - 2 | 275 | 6391053 | 77 | |
| BY790669.1 | - | 1 - 2 | 273 | 77 | ||
| Z49916.1 | - | 1 - 2 | 186 | 77 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC110668.1,Z49916.1 transcripts.
Set Membership: |
|

Clcn4-2
Genomic Map