Assay Details
| Gene Symbol: Clcn4-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035804.1 | - | 11 - 12 | 2258 | 111 | |
| AK088317.1 | - | 8 - 9 | 3644 | 111 | ||
| AK133759.1 | - | 12 - 13 | 2192 | 111 | ||
| BC005553.1 | - | 12 - 13 | 2226 | 3586154 | 111 | |
| BC110668.1 | - | 12 - 13 | 2618 | 6391053 | 111 | |
| DQ186662.1 | - | 10 - 11 | 2116 | 111 | ||
| Z49916.1 | - | 12 - 13 | 2529 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962731 | BC110668.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962731 | AK050833.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962731 | NM_011334.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538019 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670887 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716433 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341966 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 176300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79590 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88104 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105753 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101563 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962731 | BC110668.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962731 | AK050833.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962731 | NM_011334.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03805933 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517439 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03805930 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517434 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517461 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03805928 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517437 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517430 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517473 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03805921 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03805912 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101563 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03805923 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517453 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517441 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03805910 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517466 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105753 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517435 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517465 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517426 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03805924 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517452 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09517425 | - | - |
| Whole Mouse Genome G4122A | A_52_P85200 | DQ186662.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85200 | BC005553.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85200 | BC110668.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85200 | AK088317.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85200 | AK133759.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127847 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115603 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105753 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32873831 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559864 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559862 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559858 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559868 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559863 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559856 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559869 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559861 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559854 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559867 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559855 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559870 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559865 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559857 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559859 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559866 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559860 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448161_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415986_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559853 | - | - |
| Mouse Genome 430 2.0 Array | 1415986_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448161_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448161_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1415986_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12727_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12727_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700020209.mm.1 | TC0700002226.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700020209.mm.1 | NM_011334.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020210.mm.1 | TC0700002226.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700020210.mm.1 | NM_011334.4 | Refseq |
| Mouse Expression Set 430 | 1448161_a_at | - | - |
| Mouse Expression Set 430 | 1415986_PM_at | - | - |
| Mouse Expression Set 430 | 1448161_PM_a_at | - | - |
| Mouse Expression Set 430 | 1415986_at | - | - |
| Murine Genome U74v2 Set | 104704_at | - | - |
| Murine Genome U74v2 Set | 99672_PM_at | - | - |
| Murine Genome U74v2 Set | 104704_PM_at | - | - |
| Murine Genome U74v2 Set | 99672_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448161_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415986_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448161_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415986_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448161_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448161_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415986_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415986_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101500364 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3780154 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3780154 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101500364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780154 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2635086 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215984 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2635086 | - | - |
| mouse-6 v1.1 (Illumina) | scl012727.1_314-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780154 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101500364 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl012727.1_314-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101500364 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780154 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255684 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215984 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255684 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl012727.1_314-S | - | - |

Clcn4-2
Genomic Map