Assay Details
| Gene Symbol: Ccl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011338.2 | NP_035468.1 | 3 - 4 | 430 | 111 | |
| GenBank mRNA | AF128195.1 | - | 3 - 4 | 277 | 111 | |
| AF128196.1 | - | 3 - 4 | 277 | 111 | ||
| AF128197.1 | - | 3 - 4 | 277 | 111 | ||
| AF128198.1 | - | 3 - 4 | 277 | 111 | ||
| AF128199.1 | - | 3 - 4 | 277 | 111 | ||
| AF128200.1 | - | 3 - 4 | 277 | 111 | ||
| AF128201.1 | - | 3 - 4 | 277 | 111 | ||
| AF128202.1 | - | 3 - 4 | 277 | 111 | ||
| AF128203.1 | - | 3 - 4 | 277 | 111 | ||
| AF128204.1 | - | 3 - 4 | 277 | 111 | ||
| AK151131.1 | - | 3 - 4 | 298 | 111 | ||
| AK151649.1 | - | 3 - 4 | 298 | 111 | ||
| AK154511.1 | - | 3 - 4 | 298 | 111 | ||
| AK154657.1 | - | 3 - 4 | 298 | 111 | ||
| AK155032.1 | - | 3 - 4 | 298 | 111 | ||
| AK155036.1 | - | 3 - 4 | 298 | 111 | ||
| BC145962.1 | - | 3 - 4 | 282 | 40131265 | 111 | |
| BE626870.1 | - | 3 - 4 | 278 | 111 | ||
| U15209.1 | - | 3 - 4 | 411 | 111 | ||
| U19482.1 | - | 3 - 4 | 310 | 111 | ||
| U49513.1 | - | 3 - 4 | 286 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185660 | U15209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185660 | U19482.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185660 | U49513.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185660 | AK154657.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185660 | AK155032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185660 | U15209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185660 | U19482.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185660 | U49513.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185660 | AK154657.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185660 | AK155032.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691245 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704946 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89558 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121760 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185660 | U15209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185660 | U19482.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185660 | U49513.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185660 | AK154657.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185660 | AK155032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185660 | U15209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185660 | U19482.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185660 | U49513.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185660 | AK154657.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185660 | AK155032.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185660 | U15209.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185660 | U19482.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185660 | U49513.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185660 | AK154657.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185660 | AK155032.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185660 | U15209.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185660 | U19482.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185660 | U49513.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185660 | AK154657.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185660 | AK155032.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27972315 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121760 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02379821 | - | - |
| Whole Mouse Genome G4122A | A_51_P185660 | U15209.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185660 | U19482.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185660 | U49513.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185660 | AK154657.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185660 | AK155032.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185660 | U15209.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185660 | U19482.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185660 | U49513.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185660 | AK154657.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185660 | AK155032.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110634 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121760 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09390859 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09118118 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09390864 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09390858 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09390857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27972246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09034915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09241175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09118118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09457810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09650345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09650348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09390858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09611037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09457808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09650344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09390859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09611046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27972324 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10389216 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389221 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389217 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389220 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389215 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389218 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389219 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448898_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417936_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10389214 | - | - |
| Mouse Genome 430 2.0 Array | 1448898_at | - | - |
| Mouse Genome 430 2.0 Array | 1448898_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417936_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417936_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20308_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20308_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100031127.mm.1 | NM_011338.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031127.mm.1 | NM_011338.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031127.mm.1 | TC1100003397.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031125.mm.1 | NM_011338.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031125.mm.1 | NM_011338.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031125.mm.1 | TC1100003397.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031128.mm.1 | NM_011338.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031128.mm.1 | NM_011338.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031128.mm.1 | TC1100003397.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448898_at | - | - |
| Mouse Expression Set 430 | 1417936_at | - | - |
| Mouse Expression Set 430 | 1448898_PM_at | - | - |
| Mouse Expression Set 430 | 1417936_PM_at | - | - |
| Murine Genome U74v2 Set | 104388_at | - | - |
| Murine Genome U74v2 Set | 104388_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448898_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448898_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417936_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417936_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417936_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448898_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417936_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448898_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4610725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004610725 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2776603 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2776603 | - | - |
| mouse-6 v1.1 (Illumina) | scl39819.5_328-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4610725 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39819.5_328-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610725 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2776603 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2776603 | - | - |

Ccl9
Genomic Map