Assay Details
| Gene Symbol: Sema3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011349.3 | NP_035479.2 | 3 - 4 | 476 | 84 | |
| GenBank mRNA | AF036162.1 | - | 1 - 2 | 134 | 84 | |
| AF080090.1 | - | 2 - 3 | 289 | 84 | ||
| AF080091.1 | - | 2 - 3 | 275 | 84 | ||
| AK053632.1 | - | 3 - 4 | 477 | 84 | ||
| AK140340.1 | - | 3 - 4 | 928 | 84 | ||
| BC010976.1 | - | 3 - 4 | 397 | 4219060 | 84 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK172610.1 | - | 869 | Chr. - - [-] on Build | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema3f
Genomic Map