Assay Details
Gene Symbol: Sema4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF038652.1 | - | 10 - 11 | 1331 | 58 | |
AK035993.1 | - | 11 - 12 | 1604 | 58 | ||
AK047067.1 | - | 11 - 12 | 1588 | 58 | ||
AK143196.1 | - | 11 - 12 | 1616 | 58 | ||
BC057048.1 | - | 10 - 11 | 1480 | 58 | ||
BC090625.1 | - | 11 - 12 | 1572 | 58 | ||
BC094567.1 | - | 11 - 12 | 1599 | 6834138 | 58 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
