Assay Details
| Gene Symbol: Sema7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011352.2 | NP_035482.1 | 6 - 7 | 670 | 65 | |
| GenBank mRNA | AB017532.1 | - | 6 - 7 | 659 | 65 | |
| AF030699.1 | - | 6 - 7 | 666 | 65 | ||
| AF176670.1 | - | 6 - 7 | 659 | 65 | ||
| AK156123.1 | - | 6 - 7 | 693 | 65 | ||
| BC057875.1 | - | 6 - 7 | 670 | 5319572 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sema7a
Genomic Map