Assay Details
Gene Symbol: Sema7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011352.2 | NP_035482.1 | 9 - 10 | 1108 | 78 | |
GenBank mRNA | AB017532.1 | - | 9 - 10 | 1097 | 78 | |
AF030699.1 | - | 9 - 10 | 1104 | 78 | ||
AF176670.1 | - | 9 - 10 | 1097 | 78 | ||
AK156123.1 | - | 9 - 10 | 1131 | 78 | ||
BC057875.1 | - | 9 - 10 | 1108 | 5319572 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
