Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000001244.mm.1 |
NM_011361.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001244.mm.1 |
NM_011361.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001244.mm.1 |
TC1000000171.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_001161845.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_001161845.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_001161847.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_001161847.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_001161848.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_001161848.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_001161849.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_001161849.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_001161850.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_001161850.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_011361.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
NM_011361.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001246.mm.1 |
TC1000000171.mm.1 |
MTA TC |