Assay Details
| Gene Symbol: Slc16a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011391.1 | NP_035521.1 | 3 - 4 | 410 | 77 | |
| GenBank mRNA | AF058054.1 | - | 3 - 4 | 410 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF058054.1, NM_011391.1 transcripts.
Set Membership: |
|

Slc16a7
Genomic Map