Assay Details
| Gene Symbol: Slc1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077514.3 | NP_001070982.1 | 4 - 5 | 1159 | 64 | |
| NM_001077515.2 | NP_001070983.1 | 4 - 5 | 586 | 64 | ||
| NM_011393.2 | NP_035523.1 | 4 - 5 | 586 | 64 | ||
| GenBank mRNA | AB007810.1 | - | 4 - 5 | 1216 | 64 | |
| AB007811.1 | - | 4 - 5 | 575 | 64 | ||
| AB007812.1 | - | 4 - 5 | 586 | 64 | ||
| AK134609.1 | - | 4 - 5 | 1160 | 64 | ||
| AK140095.1 | - | 4 - 5 | 1154 | 64 | ||
| BC138256.1 | - | 4 - 5 | 586 | 8861275 | 64 | |
| D43796.1 | - | 4 - 5 | 739 | 64 | ||
| KC825358.1 | - | 3 - 4 | 548 | 64 | ||
| U11763.1 | - | 4 - 5 | 744 | 64 | ||
| U24699.1 | - | 3 - 4 | 557 | 64 | ||
| U75372.1 | - | 4 - 5 | 1172 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a2
Genomic Map