Assay Details
| Gene Symbol: Slc2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011400.3 | NP_035530.2 | 1 - 2 | 235 | 62 | |
| GenBank mRNA | AK089353.1 | - | 1 - 2 | 177 | 62 | |
| AK155252.1 | - | 1 - 2 | 177 | 62 | ||
| AK170425.1 | - | 1 - 2 | 177 | 62 | ||
| AK170873.1 | - | 1 - 2 | 177 | 62 | ||
| BC055340.1 | - | 1 - 2 | 204 | 6808315 | 62 | |
| BY233228.1 | - | 1 - 2 | 235 | 62 | ||
| M22998.1 | - | 1 - 2 | 139 | 62 | ||
| M23384.1 | - | 1 - 2 | 214 | 62 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK089353.1,AK155252.1,AK170425.1,AK170873.1,BC055340.1,M22998.1,M23384.1,NM_011400.3 transcripts.
Set Membership: |
|

Slc2a1
Genomic Map