Assay Details
| Gene Symbol: Slc2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011400.3 | NP_035530.2 | 3 - 4 | 483 | 67 | |
| GenBank mRNA | AK155252.1 | - | 3 - 4 | 425 | 67 | |
| AK170425.1 | - | 3 - 4 | 425 | 67 | ||
| AK170873.1 | - | 3 - 4 | 425 | 67 | ||
| BC055340.1 | - | 3 - 4 | 452 | 6808315 | 67 | |
| M22998.1 | - | 3 - 4 | 387 | 67 | ||
| M23384.1 | - | 3 - 4 | 462 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc2a1
Genomic Map