Assay Details
| Gene Symbol: Slc2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011400.3 | NP_035530.2 | 7 - 8 | 1183 | 58 | |
| GenBank mRNA | AK089353.1 | - | 7 - 8 | 1125 | 58 | |
| AK155252.1 | - | 7 - 8 | 1125 | 58 | ||
| AK170425.1 | - | 7 - 8 | 1125 | 58 | ||
| AK170873.1 | - | 7 - 8 | 1125 | 58 | ||
| BC055340.1 | - | 7 - 8 | 1152 | 6808315 | 58 | |
| M22998.1 | - | 7 - 8 | 1087 | 58 | ||
| M23384.1 | - | 7 - 8 | 1162 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc2a1
Genomic Map