Assay Details
| Gene Symbol: Slc4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011403.2 | NP_035533.1 | 14 - 15 | 2148 | 64 | |
| GenBank mRNA | AK137542.1 | - | 14 - 15 | 2044 | 64 | |
| AK157181.1 | - | 14 - 15 | 2117 | 64 | ||
| AK157833.1 | - | 14 - 15 | 2006 | 64 | ||
| AK158143.1 | - | 14 - 15 | 2149 | 64 | ||
| AK158184.1 | - | 14 - 15 | 2114 | 64 | ||
| AK158243.1 | - | 14 - 15 | 2018 | 64 | ||
| AK158453.1 | - | 14 - 15 | 1993 | 64 | ||
| AK162616.1 | - | 14 - 15 | 1993 | 64 | ||
| AK165195.1 | - | 14 - 15 | 2005 | 64 | ||
| AK165267.1 | - | 14 - 15 | 2012 | 64 | ||
| AK171662.1 | - | 2044 | 64 | |||
| AK171808.1 | - | 14 - 15 | 1982 | 64 | ||
| AK171845.1 | - | 2044 | 64 | |||
| AK172103.1 | - | 14 - 15 | 2005 | 64 | ||
| AK172316.1 | - | 14 - 15 | 1993 | 64 | ||
| AK172395.1 | - | 14 - 15 | 2044 | 64 | ||
| AK172456.1 | - | 14 - 15 | 2028 | 64 | ||
| AK172569.1 | - | 14 - 15 | 1993 | 64 | ||
| AY296129.1 | - | 14 - 15 | 1981 | 64 | ||
| BC052419.1 | - | 14 - 15 | 1988 | 5702599 | 64 | |
| BC053429.1 | - | 14 - 15 | 1979 | 6506270 | 64 | |
| M29379.1 | - | 14 - 15 | 1975 | 64 | ||
| X02677.1 | - | 14 - 15 | 1981 | 64 | ||
| X03917.1 | - | 12 - 13 | 1825 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a1
Genomic Map