Assay Details
Gene Symbol: Stxbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011503.4 | NP_035633.1 | 2 - 3 | 96 | 65 | |
GenBank mRNA | AF263345.1 | - | 2 - 3 | 103 | 65 | |
AK088364.1 | - | 2 - 3 | 118 | 65 | ||
AK133419.1 | - | 2 - 3 | 97 | 65 | ||
BC003477.1 | - | 2 - 3 | 86 | 65 | ||
BC008102.1 | - | 2 - 3 | 106 | 3966319 | 65 | |
D42068.1 | - | 2 - 3 | 88 | 65 | ||
U19520.1 | - | 2 - 3 | 110 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
