Assay Details
Gene Symbol: Stxbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011503.4 | NP_035633.1 | 12 - 13 | 1028 | 110 | |
GenBank mRNA | AF263345.1 | - | 12 - 13 | 1035 | 110 | |
AK088364.1 | - | 12 - 13 | 1050 | 110 | ||
AK133419.1 | - | 12 - 13 | 1029 | 110 | ||
AK211462.1 | - | 4 - 5 | 278 | 110 | ||
BC003477.1 | - | 12 - 13 | 1018 | 110 | ||
BC008102.1 | - | 12 - 13 | 1038 | 3966319 | 110 | |
D42068.1 | - | 12 - 13 | 1020 | 110 | ||
U19520.1 | - | 12 - 13 | 1042 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
