Assay Details
Gene Symbol: Stxbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011503.4 | NP_035633.1 | 16 - 17 | 1462 | 68 | |
GenBank mRNA | AF263345.1 | - | 16 - 17 | 1469 | 68 | |
AK088364.1 | - | 16 - 17 | 1484 | 68 | ||
AK133419.1 | - | 16 - 17 | 1463 | 68 | ||
BC003477.1 | - | 16 - 17 | 1452 | 68 | ||
BC008102.1 | - | 16 - 17 | 1472 | 3966319 | 68 | |
D42068.1 | - | 16 - 17 | 1454 | 68 | ||
U19520.1 | - | 16 - 17 | 1476 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
