Assay Details
| Gene Symbol: Tap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011530.3 | NP_035660.3 | 9 - 10 | 1788 | 82 | |
| GenBank mRNA | AK020022.1 | - | 8 - 9 | 1864 | 82 | |
| AK079667.1 | - | 9 - 10 | 1702 | 82 | ||
| AK145234.1 | - | 9 - 10 | 1747 | 82 | ||
| AK151376.1 | - | 9 - 10 | 1698 | 82 | ||
| AK151483.1 | - | 9 - 10 | 1698 | 82 | ||
| AK151657.1 | - | 9 - 10 | 1721 | 82 | ||
| AK153287.1 | - | 9 - 10 | 1698 | 82 | ||
| AK159200.1 | - | 9 - 10 | 1754 | 82 | ||
| AK161091.1 | - | 9 - 10 | 1704 | 82 | ||
| AK169645.1 | - | 6 - 7 | 996 | 82 | ||
| BC005578.1 | - | 9 - 10 | 1688 | 3968225 | 82 | |
| BC051257.1 | - | 9 - 10 | 1701 | 6530488 | 82 | |
| BC057668.1 | - | 9 - 10 | 2075 | 82 | ||
| CT010387.1 | - | 8 - 9 | 1635 | 82 | ||
| KJ650229.1 | - | 8 - 9 | 1638 | 82 | ||
| M90459.1 | - | 8 - 9 | 1635 | 82 | ||
| U60087.1 | - | 8 - 9 | 1638 | 82 | ||
| U60088.1 | - | 8 - 9 | 1638 | 82 | ||
| U60089.1 | - | 8 - 9 | 1638 | 82 | ||
| U60090.1 | - | 8 - 9 | 1638 | 82 | ||
| U60091.1 | - | 8 - 9 | 1638 | 82 | ||
| U60092.1 | - | 8 - 9 | 1635 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tap2
Genomic Map