Assay Details
| Gene Symbol: Tie1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011587.2 | NP_035717.2 | 10 - 11 | 1633 | 76 | |
| GenBank mRNA | AK051512.1 | - | 10 - 11 | 1579 | 76 | |
| AK052192.1 | - | 10 - 11 | 1553 | 76 | ||
| AK052413.1 | - | 10 - 11 | 1566 | 76 | ||
| AK161898.1 | - | 10 - 11 | 1552 | 76 | ||
| BC046452.1 | - | 10 - 11 | 1555 | 4195879 | 76 | |
| BC057004.1 | - | 10 - 11 | 1539 | 6831929 | 76 | |
| BC060182.1 | - | 10 - 11 | 1539 | 6831929 | 76 | |
| X71425.1 | - | 10 - 11 | 1534 | 76 | ||
| X73960.1 | - | 10 - 11 | 1533 | 76 | ||
| X80764.1 | - | 10 - 11 | 1847 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tie1
Genomic Map