Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000007907.mm.1 |
NM_011595.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007907.mm.1 |
NM_011595.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007907.mm.1 |
TC1000001044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007898.mm.1 |
NM_011595.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007898.mm.1 |
NM_011595.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007898.mm.1 |
TC1000001044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007900.mm.1 |
NM_011595.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007900.mm.1 |
NM_011595.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007900.mm.1 |
TC1000001044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007901.mm.1 |
NM_011595.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007901.mm.1 |
NM_011595.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007901.mm.1 |
TC1000001044.mm.1 |
MTA TC |