Assay Details
Gene Symbol: Atp6v0a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011596.5 | NP_035726.2 | 2295 | 69 | ||
GenBank mRNA | AB022323.1 | - | 2261 | 69 | ||
AF218252.1 | - | 2280 | 69 | |||
AF388674.1 | - | 4 - 5 | 469 | 69 | ||
AK017467.1 | - | 3 - 4 | 319 | 69 | ||
AK032909.1 | - | 3 - 4 | 1049 | 69 | ||
AK155055.1 | - | 2272 | 69 | |||
AK170122.1 | - | 2792 | 69 | |||
BC018369.1 | - | 2248 | 69 | |||
BC108991.1 | - | 2192 | 40047832 | 69 | ||
BC108992.1 | - | 2192 | 40047833 | 69 | ||
BC112905.1 | - | 2255 | 3670722 | 69 | ||
M31226.1 | - | 2191 | 69 | |||
X55184.1 | - | 2192 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
