Assay Details
Gene Symbol: Cd70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011617.2 | NP_035747.1 | 2 - 3 | 381 | 66 | |
GenBank mRNA | BC125407.1 | - | 2 - 3 | 340 | 40129922 | 66 |
BC125409.1 | - | 2 - 3 | 340 | 40129924 | 66 | |
U78091.1 | - | 2 - 3 | 263 | 66 | ||
Y13636.1 | - | 2 - 3 | 381 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029319 | NM_011617.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029319 | U78091.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029319 | Y13636.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262858 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930439 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147695 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029319 | NM_011617.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029319 | U78091.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029319 | Y13636.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508640 | BC125407.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508640 | BC125409.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508640 | NM_011617.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508640 | Y13636.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260275 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260256 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260258 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260260 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260242 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260239 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260240 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260253 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147695 | - | - |
Whole Mouse Genome G4122A | A_51_P508640 | BC125407.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P508640 | BC125409.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P508640 | NM_011617.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P508640 | Y13636.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101298 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147695 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13065971 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13094840 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13228958 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13582472 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13065968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260271 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13575141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260258 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260242 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260215 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260261 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260232 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260241 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260262 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260225 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260253 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260233 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260250 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13340603 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260263 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260217 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260271 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260222 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260214 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260235 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260244 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260221 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13094843 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260245 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260264 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260239 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13065971 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260265 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260257 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260240 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260267 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260229 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260249 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13147671 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260256 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13057079 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260238 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260254 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31260219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13126386 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10452305 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10452302 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10452304 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10452306 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10452303 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449926_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10452301 | - | - |
Mouse Genome 430 2.0 Array | 1449926_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1449926_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21948_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21948_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700022057.mm.1 | NM_011617.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700022057.mm.1 | NM_011617.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700022057.mm.1 | TC1700002350.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700022061.mm.1 | NM_011617.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700022061.mm.1 | NM_011617.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700022061.mm.1 | TC1700002350.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700022058.mm.1 | NM_011617.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700022058.mm.1 | NM_011617.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700022058.mm.1 | TC1700002350.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700022059.mm.1 | NM_011617.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700022059.mm.1 | NM_011617.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700022059.mm.1 | TC1700002350.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700022060.mm.1 | NM_011617.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700022060.mm.1 | NM_011617.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700022060.mm.1 | TC1700002350.mm.1 | MTA TC |
Mouse Expression Set 430 | 1449926_PM_at | - | - |
Mouse Expression Set 430 | 1449926_at | - | - |
Murine Genome U74v2 Set | 98318_at | - | - |
Murine Genome U74v2 Set | 98318_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449926_at | - | - |
Mouse Genome 430A 2.0 Array | 1449926_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449926_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449926_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 872692 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6840008 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6840008 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006840008 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2476817 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2476817 | - | - |
mouse-6 v1.1 (Illumina) | scl49744.3.1_122-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6840008 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl49744.3.1_122-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840008 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2996338 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2996338 | - | - |
