Assay Details
| Gene Symbol: Tfrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011638.4 | NP_035768.1 | 2 - 3 | 198 | 66 | |
| GenBank mRNA | AK048228.1 | - | 2 - 3 | 112 | 66 | |
| AK088961.1 | - | 2 - 3 | 112 | 66 | ||
| BC054522.1 | - | 2 - 3 | 83 | 6406097 | 66 | |
| BY101284.1 | - | 2 - 3 | 199 | 66 | ||
| X57349.1 | - | 1 - 2 | 34 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfrc
Genomic Map