Assay Details
| Gene Symbol: Tfrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011638.4 | NP_035768.1 | 14 - 15 | 1710 | 140 | |
| GenBank mRNA | AK088961.1 | - | 14 - 15 | 1624 | 140 | |
| BC054522.1 | - | 14 - 15 | 1595 | 6406097 | 140 | |
| X57349.1 | - | 13 - 14 | 1546 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfrc
Genomic Map