Assay Details
| Gene Symbol: G3bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF145285.1 | - | 3 - 4 | 332 | 105 | |
| AK076117.1 | - | 3 - 4 | 382 | 105 | ||
| AK145697.1 | - | 3 - 4 | 381 | 105 | ||
| AK149427.1 | - | 3 - 4 | 360 | 105 | ||
| AK151732.1 | - | 3 - 4 | 416 | 105 | ||
| AK151965.1 | - | 3 - 4 | 349 | 105 | ||
| AK152792.1 | - | 3 - 4 | 349 | 105 | ||
| AK153221.1 | - | 3 - 4 | 372 | 105 | ||
| AK153893.1 | - | 3 - 4 | 320 | 105 | ||
| AK167186.1 | - | 3 - 4 | 384 | 105 | ||
| AK172256.1 | - | 3 - 4 | 349 | 105 | ||
| AK193944.1 | - | 2 - 3 | 132 | 105 | ||
| AK204630.1 | - | 2 - 3 | 132 | 105 | ||
| BC048176.1 | - | 3 - 4 | 369 | 6825491 | 105 | |
| BC119806.1 | - | 3 - 4 | 250 | 40107463 | 105 | |
| BC122883.1 | - | 3 - 4 | 250 | 40107462 | 105 | |
| CN527272.1 | - | 3 - 4 | 456 | 105 | ||
| U65313.1 | - | 3 - 4 | 332 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

G3bp2
Genomic Map