Assay Details
Gene Symbol: Nek3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF093416.1 | - | 10 - 11 | 976 | 64 | |
AF099066.1 | - | 11 - 12 | 998 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
