Assay Details
Gene Symbol: Nek3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162947.1 | NP_001156419.1 | 13 - 14 | 1359 | 83 | |
NM_011848.4 | NP_035978.2 | 14 - 15 | 1411 | 83 | ||
GenBank mRNA | AF093416.1 | - | 13 - 14 | 1364 | 83 | |
AF099066.1 | - | 14 - 15 | 1380 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
