Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300010780.mm.1 |
NM_001289479.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010780.mm.1 |
TC0300001338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300010782.mm.1 |
NM_001289477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010782.mm.1 |
NM_001289478.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010782.mm.1 |
NM_001289479.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010782.mm.1 |
NM_011863.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010782.mm.1 |
NM_011863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010782.mm.1 |
TC0300001338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300010783.mm.1 |
NM_001289477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010783.mm.1 |
NM_001289478.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010783.mm.1 |
NM_001289479.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010783.mm.1 |
NM_011863.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010783.mm.1 |
NM_011863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010783.mm.1 |
TC0300001338.mm.1 |
MTA TC |