Assay Details
Gene Symbol: Slc26a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011867.3 | NP_035997.1 | 12 - 13 | 1653 | 94 | |
GenBank mRNA | AF167411.1 | - | 12 - 13 | 1654 | 94 | |
BC140995.1 | - | 11 - 12 | 1501 | 9055607 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
